Hydraulic Repair Kit (Gray only), 12 Ton, Carver
Supplier: Carver
3729
29999-824EA 735.62 CAD
29999-824
Hydraulic Repair Kit (Gray only), 12 Ton, Carver
Presses
This hydraulic repair unit contains hydraulic oil, seals, gaskets, pins, fittings, valve parts, and instructions needed to rebuild the unit.
Ordering information: Carver repair kits include all wear components that are replaceable for the 12 and 25 ton (gray only), and 30 ton presses. In most cases replacing these components will solve hydraulic unit issues. In units with excessive wear or damage to cylinders, rods, permanent valve seats and more. The repair kit may not be 100% effective. Carver repair kits are a non-refundable item.