Micromesh™ Biopsy Cassettes in Quickload™ Sleeves, Simport Scientific
Supplier: Simport Scientific
M507-2SL M507-3SL M507-4SL M507-5SL M507-6SL M507-7SL M507-8SL M507-9SL M507-10SL M507-11SL M507-12SL M507-16SL M507-17SL M507-18SL
CA87002-424CS 898.32 CAD
CA87002-424 CA87002-426 CA87002-428 CA87002-430 CA87002-432 CA87002-434 CA87002-436 CA87002-438 CA87002-440 CA87002-442 CA87002-444 CA87002-446 CA87002-448 CA87002-450
Micromesh™ Biopsy Cassettes in Quickload™ Sleeves, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
Suitable for hoppers accepting plastic sleeves, these cassettes will load in cassette labeling instruments in one simple operation. The QuickLoad™ sleeve loads directly into the hopper and is ready for printing.
  • Micromesh™ offers 0.38 mm square openings
  • Large anterior slots allow for increased fluid exchange and faster sinking in liquids
  • No biopsy pads are necessary
  • Cassettes feature a recessed cover and a large square 27 mm compartment, perfect even for needle biopsies
  • Made from Acetal
Molded from a special high density polymer, these cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of histological solvents. The lid can be opened and closed as often as necessary and always relocks securely without danger of specimen loss. Anterior writing area is at a 45° angle to make the cassette more suitable to be used with automated cassette printers.
Certifications: For IVD use.