Ziprasidone ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Ziprasidonum, 5-(2-(4-(Benzo[d]isothiazol-3-yl)piperazin-1-yl)ethyl)-6-chloroindolin-2-one

Z0034-100MG Z0034-1G
TCZ0034-100MGEA 115.69 CAD
TCZ0034-100MG TCZ0034-1G
Ziprasidone ≥98.0% (by HPLC)
Ziprasidone
Formula: C₂₁H₂₁ClN₄OS
MW: 412.94 g/mol
MDL Number: MFCD00866661
CAS Number: 146939-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa9f8fb9833b0ed0bd24ad1b5a57df4629f9a058&vid=fa9f8fb9833b0ed0bd24ad1b5a57df4629f9a058&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR