Inserts for Screw Top Vials, Basik™, 8 mm, Microsolv
Supplier: MicroSolv Technology Corp
9582S-02N 9582S-05N-K
97054-592PK 210.05 CAD
97054-592 97054-602
Inserts for Screw Top Vials, Basik™, 8 mm, Microsolv
Vials
Low volume inserts for screw top vials.
  • First hydrolytic, Class A, Type N51A, borosilicate glass
Features a precision point conical tip for excellent recovery of sample. Residual volume is <1 µl with most autosamplers with a 100 µl useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use.
Use with 8 mm, narrow mouth screw top or narrow mouth aerum finish crimp top 12×32 mm autosampler vials.
Packaging: 100/Pack.