Argos Technologies® PolarSafe® EVA Foam Ice Buckets and Pans, Cole-Parmer
Supplier: Antyila Scientific
04393-69 04393-76 04393-78 04393-81 04393-88 04393-90 04393-93 04393-54 04393-57 04393-60 04393-64 04394-02 04394-06 04394-09
76302-984EA 338.19 CAD
76302-984 76302-986 76302-988 76302-990 76302-992 76302-994 76302-996 76302-998 76303-000 76303-002 76303-004 76303-006 76303-008 76303-010 76303-012
Argos Technologies® PolarSafe® EVA Foam Ice Buckets and Pans, Cole-Parmer
Ice Containers
Mess-free superior insulation for any cryogenic medium.
  • Ethylene vinyl acetate is leak proof and will not sweat
  • Thick, nontoxic foam is compatible with a variety of cryogenic media
  • Ergonomic handles on bases and lids are glove friendly
  • Pour spouts make cleanup easy