Sodium hydroxide 0.05 M NaOH (0.05N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Caustic soda

35249-1L 35249-500ML
BJ35249-1LEA 150.57 CAD
BJ35249-1L BJ35249-500ML
Sodium hydroxide 0.05 M NaOH (0.05N) volumetric solution, Fluka™
Sodium hydroxide
Formula: NaOH
Boiling Pt: 100 °C (1.013 hPa)
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Vapor Pressure 3 mmHg ( 37 °C)
Factor 0.995 - 1.005
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR