Compound E ≥98% (by HPLC), white to off-white solid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
(S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide
02183514.2
0218351401
0218351405
IC02183514.2EA
292.05
CAD
IC02183514.2
IC0218351401
IC0218351405
Compound E ≥98% (by HPLC), white to off-white solid
Compound E
Cell permeable, potent, selective, non-transition state and non-competitive gamma-secretase inhibitor. Notch processing inhibitor. Only weakly affects presenilinase activity.
- Purity : >98% (HPLC)
- Product Type : Biochemicals
- Host : Synthetic
Solubility : Soluble in DMSO.
Formula:
C₂₇H₂₄F₂N₄O₃ Storage Temperature: Freezer |
MDL Number:
MFCD04974531 CAS Number: 209986-17-4 |
Specification Test Results
Identity Test | Passes |
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...