(+)-Catechin hydrate ≥95% (by NMR), off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: (+)-Catechin, (+)-3,3',4',5,7-Flavanpentol hydrate, (2R,3S)-2-(3,4-Dihydroxyphenyl)chromane-3,5,7-triol

02183425.5 0218342501
IC02183425.5EA 116.18 CAD
IC02183425.5 IC0218342501
(+)-Catechin hydrate ≥95% (by NMR), off-white powder
(+)-Catechin hydrate

Specific histidine decarboxylase inhibitor. Anti-inflammatory. COX-1 inhibitor. Antioxidant flavonoid. Free radical scavenger. Inhibits lipid peroxidation. Antimetastatic and anticancer compound. Antiangiogenic. Anti-osteoporotic. Antibacterial and antifungal. Neuroprotective. Mono oxidase B inhibitor. Apoptosis and cell cycle arrest inducer. JNK phosphorylation inhibitor. Antispasmodic. Insulin-mimetic.


  • Purity : >95% (NMR)
  • Product Type : Biochemicals
  • Host : Isolated from Uncaria rhynchophylla


Solubility : Soluble in DMSO, ethanol or dimethylformamide. Sparingly soluble in water.


Caution: Protect from light, store under nitrogen

Formula: C₁₅H₁₄O₆·xH₂O
MW: 290.27 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00075649
CAS Number: 154-23-4

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR