AQ™ 96 Position Vials with Caps, MicroSolv
Supplier: MicroSolv Technology Corp
95001-0AP 95001-0CP
97054-542PK 131.05 CAD
97054-542 97054-544
AQ™ 96 Position Vials with Caps, MicroSolv
Vials
These vials are designed specifically for the Waters 96 position autosamplers so their performance is assured.
  • Polyethylene caps with penetration point
These will not clog, bend or break the needles.
Packaging: Packed in AQ™ quality kit packs.