Cetrinix Supplement, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
FD029-5VL FD029-5X5VL
CA71005-624PK 485.01 CAD
CA71005-624 CA71005-626
Cetrinix Supplement, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement recommended for the selective isolation of Pseudomonas species.
  • Store at 2 to 8 °C