Dry Bath Heaters with Touch Screen
Supplier: Boekel
115001 115002 115004
76304-422EA 1191.07 CAD
76304-422 76304-426 76304-504
Dry Bath Heaters with Touch Screen
Block Heaters Block Heater/Coolers
Boekel scientific's dry block heaters are advanced, configurable and easy-to-use.
  • Easily viewable touch screen
  • Save up to 25 programs for rapid sample processing
  • Slim design to conserve bench space
  • Wide temperature range to fit many applications
  • Settable timer up to 99 hours or continuous
The Boekel scientific block heaters line is designed to accommodate the most basic sample incubation procedures as well as multi-step processes and thermal cycling procedures. The slim one, two and four block design allows for more bench space and a configurable workMore Product Information