Rose Bengal Chloramphenicol Agar, HiMedia

Supplier: HIMEDIA LABORATORIES PVT. CA

M640-500G M640-2.5KG M640-5KG
CA95022-944EA 1320.6 CAD
CA95022-944 CA89226-112 CA89226-114
Rose Bengal Chloramphenicol Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology

Rose bengal chloramphenicol agar is used for selective isolation and enumeration of yeasts and moulds from foods and environmental materials.


  • Appearance: Light yellow to pink homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.55% agar gel
  • Colour and clarity of prepared medium: Deep pink colored clear to slightly opalescent gel forms in petri plates
  • Reaction: Reaction of 3.22% w/v aqueous solution at 25 °C, pH: 7.2 ±0.2


Mycological peptone or papaic digest of soyabean meal provides essential growth nutrients. Dextrose is the fermentable carbohydrate. Chloramphenicol has inhibitory action on gram-negative bacteria. Rose bengal dye suppresses the development of bacteria and reduces the spreading of moulds, controls the size and height of mould colonies such as Rhizopus species. Rose bengal is taken up by mould and yeast colonies thereby assist in enumeration.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR