Protector® Evidence Drying Cabinets with Washdown, Labconco
Supplier: Labconco
3400010 3400011 3400012 3400013 3400014
89412-208EA 15468.2 CAD
89412-208 89412-210 89412-212 89412-214 89412-216
Protector® Evidence Drying Cabinets with Washdown, Labconco
Fume Hoods and Enclosures Forensic Enclosures
The durable, stainless steel-lined Protector Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding. The cabinet’s HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The spray nozzle allows the interior to be washed down for easy clean up after the cabinet has been properly disinfected. A built-in water pump aids clean up.
  • Built-in blower with solid state speed control
  • Epoxy-coated steel exterior
  • Red, filter system alarm light to alert operator to
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable