LB Agar Plates, Glucose 1%, Kanamycin-100, Teknova
Supplier: Teknova
L1827 L1827
CA100216-624PK 841.93 CAD
CA100216-624 100216-624
LB Agar Plates, Glucose 1%, Kanamycin-100, Teknova
Media for Microbiology Prepared Media for Microbiology
Ready to use LB agar plates (miller formulation) for the growth of recombinant E. Coli. All plates are made with the highest quality products and tested with four strains of E. coli.
Formulation includes 1% tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1% glucose, 100 µg/ml kanamycin and 1.5% agar.