Wireless Humidity/Temperature Monitor Set
Supplier: Sper Scientific
800255C 800255 800254 800254C
10852-656EA 121.27 CAD
10852-656 10852-670 10852-724 10852-726
Wireless Humidity/Temperature Monitor Set
Monitoring Systems CO2 and Temperature Monitoring Systems
Monitor RH and temperature inside an environmental chamber or clean room without opening the door. Place the weatherproof (IPX3) sensor outdoors, in green houses, warehouses, air ducts and other hard to reach places and continuously monitor ambient RH and temperature from a remote location. The wireless sensors broadcast to the monitor from up to 100 feet (30 m) away without obstructions, using a wireless RF signal.
The monitor displays the local and remote temperatures in ºC or ºF, recalls min/max on both, and indicates when the batteries are low in both the sensors and monitor. It alsoMore Product Information