Protexis™ PI Blue with Neu-Thera® Surgical Gloves, Cardinal
Supplier: Cardinal Health
2D73EB60 2D73EB65 2D73EB70 2D73EB75 2D73EB80 2D73EB85 2D73EB90 2D73EB55
76265-658BX 732.75 CAD
76265-658 76265-660 76265-662 76265-664 76265-666 76265-668 76265-670 76265-840
Protexis™ PI Blue with Neu-Thera® Surgical Gloves, Cardinal
Gloves
These sterile polyisoprene surgical gloves with Neu-Thera® Emollient Coating can be worn as a foundational layer when double-gloving. The blue color alerts wearers to breaches in the outer glove.
  • Synthetic polyisoprene (PI) – not made from natural rubber latex
  • Distinct blue color aids in alerting wearers to perforations in the outer glove
  • Neu-Thera® Emollient Coating aids in wet and dry donning, and helps promote skin moisturization
  • Proprietary hand mold with an independent thumb design allows for an anatomical fit and natural movement in the fingers,
More Product Information