Fi-Streem™ 2S Still 20 Liter Carboy, Labstrong Corporation
Supplier: LabStrong
FI-Streem®
B00123
CA10809-190EA 3485.09 CAD
CA10809-190
Fi-Streem™ 2S Still 20 Liter Carboy, Labstrong Corporation
Bottles Carboys
The LabStrong 20 L carboy allows for fully automatic operation when connected to Fi-Streem™ 2S stills.
  • Rectangular base that occupies less space than a round carboy
  • Large opening at the top providing for easier filling and cleaning
  • Molded carboy size identification
  • Large, clear graduation marks with ±5% accuracy
  • Autoclavable dispense spigot
The ergonomically designed carboy body is coupled with a unique cap technology that provides the user with an easy-to-use and easy-to-clean product. Autoclavable lid is modified to accept the Fi-Streem™ float switch.
It's body includes design features that allow the user to easily hold and manipulate during lifting, filling and pouring. Its features include a set of pinched handles at the top which prevents the possibility of fluid trapping voids, grips at the bottom and easy-to-read identification marks. This product operates at 65 lbs.
Accessories information: The leak-proof lid for the 20 L carboys is stackable and autoclavable. The lid also features large handle grips that allow for easy tightening and opening of the cap. The cap is made from autoclavable polypropylene.