Sodium thiosulfate 0.1 M Na2S2O3 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

35245-10L-VP 35245-6X1L 35245-5L-VP
BJ35245-10L-VPEA 503.86 CAD
BJ35245-10L-VP BJ35245-1L BJ35245-5L-VP
Sodium thiosulfate 0.1 M Na2S2O3 (0.1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
Sodium thiosulphate
Formula: Na₂S₂O₃
Boiling Pt: 100 °C (1.013 hPa)
Density: 1.0 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR