Particulate Matter on Filter Paper, ERA
Supplier: ERA
1150 1050
CA10066-112EA 596.09 CAD
CA10066-112 CA10066-114
Particulate Matter on Filter Paper, ERA
Filters Filter Papers Quantitative Filter Paper
Standard filter paper for proficiency test. Particulate Matter on Filter Paper is usuable with the following methods: EPA 5, EPA 5A, EPA 5B, EPA 5D, and EPA 5F.
Particulate matter = 50 to 600 mg filter.