Skim Milk Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences
580S-9
89001-514CS 61.7 CAD
89001-514
Skim Milk Pipettes, DWK Life Sciences
Pipettes Milk Testing Pipettes
Sealed-to-contain 9 ml skim milk pipettes are designed to comply with the Pennsylvania modified Babcock method, as referenced in APHA 15.8, for testing skim milk, low-fat milk, buttermilk or whey.
  • Calibrated to contain
  • Scale is permanent brown stain
  • Specially designed rubber washer is supplied with each pipette and is used to support the pipette in the neck of the test bottle
  • Marked with sealed three for use in states requiring this marking
Pipettes are manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Material conforms to USP Type I and ASTM E438, Type 1I, Class A requirements.