ValueWare® Reusable Soda Lime Glass Petri Dishes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
53061-6015 53061-10010 53061-10015 53061-10020
76175-260CS 527.82 CAD
76175-260 76175-262 76175-264 76175-266
ValueWare® Reusable Soda Lime Glass Petri Dishes, DWK Life Sciences
Culture Dishes Petri Dishes
These shallow glass dishes are manufactured from soda lime glass.
Sold in sets consisting of top and bottom dishes.
it is autoclavable at 121 °C and can withstand temperatures up to 500 °C.