Single Scale Graduated Cylinders
Supplier: Wards
CH0346A CH0346B CH0346C CH0346D CH0346E
470310-990EA 7.45 CAD
470310-990 470310-992 470310-994 470310-996 470310-998
Single Scale Graduated Cylinders
Cylinders Measuring Cylinders
Single Scale Graduated Cylinders with plastic base and protection collar.
  • Class B
  • Borosilicate glass gradauted cylinder
  • Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.