TPX Beakers
Supplier: Wards
CH0138BPR CH0138CPR CH0138DPR CH0138APR CH0138EPR CH0138FPR CH0138GPR CH0138PRSET
470310-652EA 22.05 CAD
470310-652 470310-654 470310-656 470310-706 470310-776 470310-778 470310-780 470310-782
TPX Beakers
Beakers
These TPX (PMP) beakers are idea for general laboraotry use.
  • Chemically resistant
  • Autoclavable
  • Long lasting and high clarity beakers
Able to withstand temperatures up to 180 °C.