Glass Powder Funnels
Supplier: Wards
CH0459A CH0459B CH0459C CH0459E CH0459F
470310-692EA 6.3 CAD
470310-692 470310-694 470310-696 470310-698 470310-700
Glass Powder Funnels
Funnels Powder Funnels
These funnels are available in a variety of different diameter openings.
  • Borosilicate glass
  • Plain 60° angle
  • Fast and efficient
When pouring large particles into reaction vessels and beakers, powder funnels allow a clean and quick procedure. Powder bridging is minimized by a parallel, short stem.
Order Now
SPECIFICATIONS
Environmentally Preferable