Neoprene Stoppers
Supplier: Wards
CH0321C2H CH0321D CH0321D1H CH0321D2H CH0321F1H CH0321F2H CH0321G CH0321G1H CH0321G2H CH0321H CH0321H1H CH0321H2H CH0321I CH0321I1H CH0321I2H CH0321J CH0321J1H CH0321J2H CH0321K CH0321K1H CH0321K2H CH0321M1H CH0321M2H CH0321N CH0321N2H CH0321P1H CH0321P2H CH0321Q1H CH0321Y2H CH0321Z CH0321Z1H CH0321Z2H CH0321S CH0321S1H CH0321S2H CH0321T CH0321T1H CH0321T2H CH0321U CH0321U1H CH0321U2H CH0321W1H CH0321W2H CH0321X CH0321X1H CH0321B CH0321B1H CH0321C CH0321C1H CH0321E CH0321E1H CH0321E2H CH0321F CH0321Q2H CH0321V CH0321V1H CH0321V2H CH0321W CH0321X2H CH0321Y CH0321Y1H CH0321L CH0321L1H CH0321L2H CH0321M CH0321O CH0321O1H CH0321O2H CH0321P CH0321R CH0321R1H CH0321R2H CH0321A CH0321A1H CH0321Q CH0321N1H
470310-570PK 13.6 CAD
470310-570 470310-572 470310-574 470310-576 470310-578 470310-580 470310-582 470310-584 470310-586 470310-588 470310-590 470310-592 470310-594 470310-596 470310-598 470310-600 470310-602 470310-604 470310-606 470310-608 470310-610 470310-612 470310-614 470310-616 470310-618 470310-620 470310-622 470310-624 470310-626 470310-628 470310-630 470310-632 470310-658 470310-660 470310-662 470310-664 470310-666 470310-668 470310-670 470310-672 470310-674 470310-676 470310-678 470310-680 470310-682 470310-744 470310-746 470310-748 470310-750 470310-752 470310-754 470310-756 470310-758 470310-760 470310-762 470310-764 470310-766 470310-768 470310-770 470310-772 470310-774 470310-786 470310-788 470310-790 470310-792 470310-794 470310-796 470310-798 470310-800 470310-802 470310-804 470310-806 470310-808 470310-810 470311-000 470311-002
Neoprene Stoppers
Stoppers Rubber Stoppers
Versatile neoprene stoppers perfect for every laboratory.
  • Solid, one hole or two hole stoppers are available
  • Manufacturered according to sizes recommended by ASTM standards
Manufactured from long lasting neoprene rubbers. Suitable for use with petroleum products, mineral oils, fats as well as most inorganic acids and bases. Neoprene rubber is oil resistant and suitable for temperatures up to 120 °C (250 °F).
Packaging: Packs of 10.