Chem-Vac™ Universal Stirrer Glass Ball Bearings, Chemglass
Supplier: Chemglass
CG-2077-U-01 CG-2077-U-02 CG-2077-U-50
30622-654EA 1529.43 CAD
30622-654 30622-656 30622-658
Chem-Vac™ Universal Stirrer Glass Ball Bearings, Chemglass
Stirrers Stirring Tools
These universal Chem-Vac™ PTFE stirrer bearings provides a vacuum tight flake-free seal on PTFE, glass or stainless-steel stir shafts. Loosening nut above the standard taper inner joint allows easy and quick removal.
  • Flake-free
  • Chemically resistant
  • The polymer blend sleeve provides a long-lasting seal for use up to 500 rpm
  • For use with polished glass, PTFE or stainless-steel stirrer shafts
The upper portion of bearing rotates with the stirrer shaft providing a flake-free seal. The bearing consists of a PTFE standard taper body, glass filledMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable