KONTES® Separatory Funnels, Squibb, with PTFE Plug Stopcock, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
298290-0125 298290-0060
KT298290-0125EA 349.71 CAD
KT298290-0125 KT298290-0060
KONTES® Separatory Funnels, Squibb, with PTFE Plug Stopcock, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Squibb separatory funnel with heavy duty drip joint at the bottom.
  • Pear-shaped
  • Standard Taper 19/22 joints
  • PTFE stopcock plug
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.