Potassium tellurite concentrate for dilution to 1%, Himedia®

Supplier: HIMEDIA LABORATORIES PVT. CA

FD052-5VL FD052-5X5VL
CA71005-672PK 181.19 CAD
CA71005-672 CA71005-674
Potassium tellurite concentrate for dilution to 1%, Himedia®
di-Potassium tellurium trioxide

Recommended for the selective isolation of Staphylococci and Corynebacteria.


To achieve 1% solution dilute the contents in 8.9 ml sterile distilled water.

Formula: K₂TeO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00011379
CAS Number: 7790-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR