Perchloric acid 70% ACS
Supplier: GFS CHEMICALS INC
|
Danger
|
55504
71002-064CS
1930.41
CAD
71002-064
Perchloric acid 70% ACS
Perchloric acid
Formula:
HClO₄ MW: 100.46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 UN: 1873 ADR: 5.1(8),I |
Specification Test Results
Assay | 69.0 - 72.0% |
Residue after ignition | Max. 0.003% |
ilicate + phosphate | Max. 0.0005% |
Chloride | Max. 0.001% |
Sulfate | Max. 0.001% |
Nitrogen Compounds (as N) | Max. 0.001% |
Heavy metals (as Pb) | Max. 0.0001% |
Iron | Max. 0.0001% |
Color (APHA) | Max. 10 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...