Silicon dioxide 99,0-100,5 calcined_KT, colloidal dispersion, EMPROVE® ESSENTIAL E551, NF, JP, Ph. Eur.

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

1.13126.0500 1.13126.5000 1.13126.0500 1.13126.5000
CA1.13126.0500CS 1783.27 CAD
CA1.13126.0500 CA1.13126.5000 EM1.13126.0500 EM1.13126.5000
Silicon dioxide 99,0-100,5 calcined_KT, colloidal dispersion, EMPROVE® ESSENTIAL E551, NF, JP, Ph. Eur.
Silicium dioxide

  • Colloidal
  • Highly dispersed


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.


Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Certifications: Corresponds to Ph Eur, NF, JP, E551

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1719
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now

Specification Test Results

Identity Passes Test
Assay (Calculated on Calcined Substance) 99.0 - 100.5%
Substances Soluble in Water ≤5%
pH-value (4%, water) 3.5 - 5.5
Chloride (Cl) ≤0.011%
Heavy Metals (as Pb) ≤0.0025%
Al (Aluminium) ≤0.0010%
As (Arsenic) ≤0.0003%
Ca (Calcium) Conforms
Fe (Iron) ≤0.0500%
Hg (Mercury) ≤0.0001%
Pb (Lead) ≤0.0005%
Pb (Lead) (ICP) ≤0.0005%
Residual solvents (ICH Q3C) Excluded by Production Process
Loss on Ignition (Dried Substance) ≤2.0%
Loss on Drying ≤2.5%
Spec. Surface 200 ± 25
Volume Test Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR