High Pressure Sample Pans, PerkinElmer
Supplier: PerkinElmer
B0182902 03190029 B0182901
CA97012-954EA 3569.54 CAD
CA97012-954 CA97013-690 CA97013-784
High Pressure Sample Pans, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans contain the sample in a sealed environment throughout an experiment.
  • Pressure ranges from 24 to 150 bar
The sealing suppresses the vaporization of a solvent or contains a volatile reaction product, thereby eliminating the interfering effects of the heat of vaporization.
The stainless steel pans with covers and seals are reusable and includes 5 pans and 20 gold-plated seals.