Stirrer Bearings, PTFE, Chemglass
Supplier: Chemglass
CG-2077-H-06 CG-2077-H-12 CG-2077-H-13 CG-2077-H-14 CG-2077-H-01 CG-2077-H-02 CG-2077-H-07 CG-2077-H-10 CG-2077-H-11 CG-2077-H-20 CG-2077-H-21 CG-2077-H-22 CG-2077-H-23 CG-2077-H-30 CG-2077-H-31 CG-2077-H-32 CG-2077-H-33 CG-2077-H-40 CG-2077-H-41 CG-2077-H-42 CG-2077-H-43
33501-550EA 1310.67 CAD
33501-550 33501-558 33501-560 33501-562 33501-544 33501-546 33501-552 33501-554 33501-556 33501-564 33501-566 33501-568 33501-570 33501-572 33501-574 33501-576 33501-578 33501-580 33501-582 33501-584 33501-586
Stirrer Bearings, PTFE, Chemglass
Stirrers Stirring Tools
These grease-free PTFE stirrer bearings are designed for use under moderate vacuum. These bearings consist of a PTFE standard taper body, nylon compression cap, PTFE insert/bushing and a nylon loosening nut.
  • PTFE insert/bushing provides glass and PTFE contact with the reaction
The loosening nut present above the standard taper inner joint allows easy and quick removal.
Delivery information: Supplied complete with a perfluoro O-ring on the body and a viton O-ring on the bushing.