Dehydrated Isolation Medium for Iron Bacteria, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M622-500G M622-2.5KG M622-5KG
CA61000-130EA 1196.27 CAD
CA61000-130 CA89226-052 CA89226-054
Dehydrated Isolation Medium for Iron Bacteria, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Isolation medium for iron bacteria is used for the isolation of iron bacteria, especially those belonging to Sphaerotilus-Leptothrix group.
  • Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C