Fiberboard Storage Boxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
1950878 1950879 1950881 1950882 1950883 1950884
CA10141-560PK 506.9 CAD
CA10141-560 CA10141-562 CA10141-566 CA10141-568 CA10141-570 CA10141-572
Fiberboard Storage Boxes, Thermo Scientific
Boxes Cryoboxes
Store a variety of vials and tubes with these water-repellant, fiberboard storage boxes.
  • Sturdy and water-repellant
  • Covers for sample storage
Ideal for use in ultra-low temperature or cryogenic freezers.
Ordering information: Choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.
Packaging: 12/Pack.