Vial Holders for HG-600 Geno/Grinder® 2010 Homogenizer
Supplier: SPEX
2196-16-PE 2197 2650 2660 2661 2662 2663 2664 2665 2666 2191 2193 2260 2300 2198 2198-24-PE 2257 2259 2014
76181-818EA 126.29 CAD
76181-818 76181-820 76181-834 76181-838 76181-840 76181-842 76181-844 76181-846 76181-848 76181-850 76182-212 76182-262 76182-314 76182-316 76533-096 76533-098 76533-100 76533-102 77391-964
Vial Holders for HG-600 Geno/Grinder® 2010 Homogenizer
Homogenizers Tissue Grinders
Securely hold vials in place while processing samples.
  • Safely grind samples in the Cole-Parmer HG-600 Geno/Grinder® 2010 Tissue Homogenizer and Cell Lyser
Vial holders are suitable for use with the Cole-Parmer HG-600 Geno/Grinder® 2010 Tissue Homogenizer and Cell Lyser. Order homogenizer and vials separately. Chill cryo-blocks in the cryo-station 2600 to maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis. The blocks preserve pesticides, RNA, and proteins for heat sensitive extraction methods.