N-Boc-aniline 97%
Supplier: Thermo Scientific Chemicals
H55232-06
H55232-14
CAAAH55232-06EA
70.15
CAD
CAAAH55232-06
CAAAH55232-14
N-Boc-aniline 97%
N-Boc aniline
Formula:
C₁₁H₁₅NO₂ Melting Pt: 133…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043723 CAS Number: 3422-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...