Palladium 10% on carbon Type 487 wetted_50%_Water

Supplier: Thermo Scientific Chemicals
Warning

H34200-09 H34200-18
CAAAH34200-09EA 522.98 CAD
CAAAH34200-09 CAAAH34200-18
Palladium 10% on carbon Type 487 wetted_50%_Water
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR