Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) ≥98% (by HPLC)
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
Cytidine-5'-triphosphate disodium, Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate
00095-250MG
00095-1G
CH00095-250MGEA
31.92
CAD
CH00095-250MG
CH00095-1G
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) ≥98% (by HPLC)
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)
Formula:
C₉H₁₄N₃Na₂O₁₄P₃ MW: 527.12 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00078193 CAS Number: 36051-68-0 |
Specification Test Results
Appearance | White powder |
Solubility | Clear colorless solution (50 mg/mL in Water) |
Water Content (Karl Fisher) | ≤12.0% |
UV Ratios, A250/A260 at pH 7.0 | 0.81 - 0.87 |
UV Ratios, A280/A260 at pH 7.0 | 0.94 - 1.00 |
Purity (HPLC) | ≥98% |
Purity (UV) | ≥96% (on Dry Basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...