Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production
Supplier: MilliporeSigma
1.37028.2500
1.37028.9010
1.37028.1000
EM1.37028.2500EA
401.15
CAD
EM1.37028.2500
EM1.37028.9010
EM1.37028.1000
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production
Glycerine
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 290 °C (1013 hPa) Melting Pt: 18.6 °C Density: 1.26 g/cm³ (20 °C) Flash Pt: 199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...