Carbenicillin disodium salt, ≥ 770 mcg/mg USP

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: NSC 111071, α-Carboxybenzylpenicillin disodium salt, Sodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na

00049-1G 00049-25G 00049-5G
CH00049-1GEA 31.92 CAD
CH00049-1G CH00049-25G CH00049-5G
Carbenicillin disodium salt, ≥ 770 mcg/mg USP
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now

Specification Test Results

Appearance White powder
Solubility (0.1% aqueous solution at RT using frequent agitation) Clear, colorless solution
Water Content (Karl Fisher) ≤6.0%
pH 6.5 - 8.0
Infrared Spectrum Conforms to Structure
Potency (Anhydrous Basis) ≥770 mcg/mg
Pyrogen Conforms to USP
Identification Test for Sodium Passes Test
Residual Solvents Conforms to USP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR