Ammonium formate ≥98% (by titrimetric analysis), white crystalline powder

00327-1KG 00327-250G 00327-5KG
CH00327-1KGEA 94.8 CAD
CH00327-1KG CH00327-250G CH00327-5KG
Ammonium formate ≥98% (by titrimetric analysis), white crystalline powder
Ammonium formate
Formula: HCO₂NH₄
Melting Pt: 119…121 °C
Density: 1.26 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2

Order Now

Specification Test Results

Appearance White crystalline powder
Assay by Titration ≥98%
Anion (Chloride (Cl⁻)) ≤5 ppm
Anion (Sulfate (SO₄²⁻)) ≤0.005%
pH 6.5 - 7.0
Insoluble ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR