RBC Lysis Buffer, IBI Scientific
Supplier: IBI Scientific
IB47620
IB47621
IB47622
89135-030EA
65.14
CAD
89135-030
89135-032
89135-034
RBC Lysis Buffer, IBI Scientific
Buffers
Extraction Buffers
RBC Lysis Buffer is used to lyse red blood cells in nucleic acid isolation applications.
- Lyses red blood cells for DNA and RNA extractions
Can be used with the Genomic DNA (Blood) Extraction Kits and the Total RNA (Blood) Extraction Kits.