LT300 Protective Eyewear, HexArmor
Supplier: HexArmor
12-10001-02 12-10002-04 12-10003-02 12-10007-05
76207-834EA 72.13 CAD
76207-834 76207-836 76207-838 76207-842 76318-178
LT300 Protective Eyewear, HexArmor
Safety Glasses
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.100% UV protection, metal-free.
  • Panoramic lens for wide field of vision
  • Fits over most prescription glasses
  • Wide, fully adjustable high-quality head strap offers pressure-free comfort
  • Compatible with other PPE, such as respirators
  • Indirect ventilation system
  • LTX™ Lens technology for premium optical clarity
  • D3/D4 Liquid splash rated
The LT300 goggle. Featuring a durable, complete coverage soft seal with an indirect channel ventilation system that safeguards against liquids, dirt, and debris.Combines soft and hard components which adapt to wearer’s face, creating a soft seal.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.
Certifications: Meets standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.