VS300 Protective Eyewear, HexArmor
Supplier: HexArmor
11-19001-02 11-19010-08 11-19002-03 11-19004-08 11-19005-09 11-19007-02 11-19009-10
76207-828EA 46.85 CAD
76207-828 76207-830 76208-240 76208-242 76208-244 76208-440 76208-442 76208-444 76208-446 76208-448
VS300 Protective Eyewear, HexArmor
Safety Glasses
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.100% UV protection, metal-free
  • Soft ear pieces guarantee a stable and low pressure fit
  • Brow guard provides protection from flying particles, dirt, and dust
  • Flexible side arms with seven inclination levels
  • Unique dual-injection hard and soft components
  • Soft flex nose loops ensure even weight distribution on nose bridge
  • LTX™ Lens Technology for premium optical clarity
The innovative VS300 with permanent anti-fog coatings. The VS300 is the luxury vehicle of safety eyewear. These premium safety glasses feature seven side arm inclination levels, allowing for a comfortable fit no matter the wearer's head shape.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.
Certifications: Meets Standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.