Ionomycin free acid ≥98% (by HPLC, TLC), waxy colorless solid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Antibiotic. Potent and highly selective Ca2+ ionophore. Commonly used to modify intracellular calcium levels to study calcium transport across biological membranes and to calibrate fluorescent Ca2+ indicators. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations at efficiencies that are greater than or equal to those for Ca2+. Apoptosis inducer. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase and induces central demyelination. Used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2 and IL-4 usually in conjunction with PMA. ADAM10 agonist. Potent inducer of shedding CXCL16. TREK-1 channels inhibitor. PPARgamma ligand with a unique binding mode. Shows effective glucose-lowering activity in a mouse model of diabetes.
- Purity : >98% (HPLC, TLC)
- Product Type : Biochemicals
- Host : Isolated from Streptomyces conglobatus.
- Solubility : Soluble in methanol or DMSO.
Formula:
C₄₁H₇₂O₉ Storage Temperature: Freezer |
MDL Number:
MFCD06798385 CAS Number: 56092-81-0 |
Specification Test Results
Identity Test | Passes |
Purity | >98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...