3-Methyladenine
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine
0215545925
0215545980
0215545983
IC0215545925EA
109.53
CAD
IC0215545925
IC0215545980
IC0215545983
3-Methyladenine
3-Methyladenine
- Product Type : Biochemicals
- Physical Description: White, Free Flowing Powder
Formula:
C₆H₇N₅ Storage Temperature: Ambient |
MDL Number:
MFCD00010531 CAS Number: 5142-23-4 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...