20-Hydroxyecdysone ≥97%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: β-Ecdysone

0215102405 0215102410 0215102450
IC0215102405EA 307.39 CAD
IC0215102405 IC0215102410 IC0215102450
20-Hydroxyecdysone ≥97%
20-Hydroxyecdysone

  • White Powder
  • Store Under Nitrogen

Formula: C₂₇H₄₄O₇
Melting Pt: 245…247 °C
Storage Temperature: Freezer
MDL Number: MFCD00036740
CAS Number: 5289-74-7

Order Now

Specification Test Results

Identity Test Passes
Purity 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR