Guinea Pig Complement, MP Biomedicals
Supplier: MP Biomedicals
0855852 0855854
IC0855852EA 94.91 CAD
IC0855852 IC0855854
Guinea Pig Complement, MP Biomedicals
Sera (Serum)
Product is guinea pig serum, rapidly processed to preserve complement activity.
  • Guinea Pig Complement is suitable for use in hemolytic titrations and cytotoxicity assays, and as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques.
  • This product has not been absorbed, and should be tested for use in specific cytotoxicity applications.
The product gives a spectrum of normal serum precipitin arcs when tested by immunoelectrophoresis with anti-guinea pig serum. The product is tested for its ability to lyse red blood cells.
Pooled guinea pig serum is filtered through a 0.22 µm filter, vialed and lyophilized. No preservative is added.
Reconstituition instructions: Reconstitute product with 1 mL of deionized or distilled water. Gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material