Human beta-2-Microglobulin (from Urine), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
08770951
08770952
08770953
IC08770951EA
103.02
CAD
IC08770951
IC08770952
IC08770953
Human beta-2-Microglobulin (from Urine), MP Biomedicals
Proteins and Peptides
Lyophilized from 0.02M Ammonium Bicarbonate (may contain traces of buffer salt).
Immunological identity verified by radial immunodiffusion. Cellulose acetate electrophoresis shows one band indicating β2 electrophoretic mobility. SDS-PAGE shows one band corresponding to a molecular weight of approximately 12,000.
Reconstitution: To reconstitute use a phosphate buffer, pH >7.0, containing 0.15 M NaCl.
- Calibrators in enzyme assays
- Cancer and renal biomarker