Safety Labels and Tags, White, BradyJet™ J2000, Brady
Supplier: Brady Worldwide
J20-260-7425J J20-261-7425J J20-262-7425J J20-18-7425J J20-18-2595 J20C-2250-7425J J20C-1125-7425J
76309-218RL 415.49 CAD
76309-218 76309-220 76309-222 76309-224 76309-226 76309-228 76309-230
Safety Labels and Tags, White, BradyJet™ J2000, Brady
Labels
Ideal for full-color identification and barcode applications, complete identification for a variety of areas through your laboratory including: cryovials, tubes, eppendorf tubes, conical, lab glassware, lab instructions or housekeeping and lab equipment markings.
  • White color
  • Matte finish
  • Ink jet print technology
B-7425J - matte white polypropylene with acrylic adhesive is inkjet printable. Perfect for laboratory identification such as vials, centrifuge tubes and test tubes. Service temperature of −112 to 158 °F (−80 to 70 °C).
B-2595 - general purpose vinyl label and B-2569 Polyester with permanent adhesive label. Specially formulated topcoat enhances colors and label appearance. Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting. Topcoat protects print from smudging and harsh conditions. Inkjet-receptive labels for use with the BradyJet™ J2000 color label printer.
B-2551 - Ideal for use in general facility labeling, including accident prevention and confined space tags. Compliant with OSHA standard 1910.145 for accident prevention tags and 1910.147 for lockout tagout.